Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-21 Antibody (461804), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB25372MFV500
Description
FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| FGF-21 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 26291 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 461804 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| FGF-21, fibroblast growth factor 21 | |
| E. coli-derived recombinant human FGF-21, His29-Ser209, Accession # Q9NSA1 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction