Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-21 Antibody (CL6491), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | FGF-21 |
---|---|
Clone | CL6491 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
FGF-21 Monoclonal specifically detects FGF-21 in Human samples. It is validated for Western Blot.Specifications
FGF-21 | |
Western Blot | |
Unconjugated | |
Human | |
FGF-21, fibroblast growth factor 21 | |
FGF21 | |
IgG1 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
CL6491 | |
Monoclonal | |
Mouse | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
26291 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: PEPPGILAPQPPDVGSSDPLSMVGPSQGR | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title