Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-21 Antibody (OTI2F10), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271815FR
Description
FGF-21 Monoclonal specifically detects FGF-21 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
FGF-21 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
FGF21 | |
Recombinant protein fragment corresponding to amino acids 29-209 of human FGF-21 (NP_061986) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2F10 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
FGF-21, fibroblast growth factor 21 | |
Mouse | |
Affinity Purified | |
Angiogenesis, Biologically Active Proteins, Cancer | |
26291.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction