Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-22 Antibody (MM0286-4F28), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212300MFV610
Description
FGF-22 Monoclonal antibody specifically detects FGF-22 in Human samples. It is validated for Western Blot, NeutralizationSpecifications
FGF-22 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Neutralization | |
MM0286-4F28 | |
Western Blot, Block/Neutralize | |
FGF-22, fibroblast growth factor 22 | |
Human recombinant FGF-22 | |
0.1 mL | |
Angiogenesis | |
27006 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction