Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-23 Antibody (FGF23/132), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320358100UG
Description
FGF-23 Monoclonal antibody specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FGF-23 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
Recombinant full-length human FGF-23 protein | |
100 μg | |
Angiogenesis, Biologically Active Proteins, Cancer | |
8074 | |
Store at 4&drg;C. Do not freeze. | |
IgG |
Immunohistochemistry (Paraffin) | |
FGF23/132 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction