Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-3 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | FGF-3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FGF-3 Polyclonal antibody specifically detects FGF-3 in Rat samples. It is validated for Western BlotSpecifications
FGF-3 | |
Polyclonal | |
Purified | |
RUO | |
Rat | |
fibroblast growth factor 3, fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog), HBGF-3, mouse, oncogene INT2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FGF-3 (NP_005238.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Tumor Suppressors | |
PBS, 50% glycerol, pH7.3 | |
2248 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title