Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF 4 Polyclonal Antibody, Abbomax

Catalog No. 502203940
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3940 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3940 Supplier Abbomax Inc Supplier No. 600890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Recombinant protein FGF-4 Cellular location: Secreted.

FGF 4 Polyclonal antibody specifically detects FGF 4 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen FGF 4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Fgf4
Gene Accession No. P08620, P11403
Gene Alias FGF; FGF4; Fgf-4; Fgfk; Fibroblast growth factor; fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; heparin secretory-transforming protein 1; heparin-binding growth factor 1; heparin-binding growth factor 4; HST; Hst1; hst-1; HSTF1; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; kaposi sarcoma oncogene; Kfgf; K-FGF; K-fibroblast growth factor; KS3; oncogene HST; transforming protein KS3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of Fibroblast Growth Factor 4 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116499, 14175, 2249
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.