Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-5 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP197760
Description
FGF-5 Polyclonal specifically detects FGF-5 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
FGF-5 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FGF-5, fibroblast growth factor 5, HBGF-5, Heparin-binding growth factor 5, Smag-82 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize reported isoform 1 (NP_004455.2) only. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.1 - 0.3 ug/mL, Peptide ELISA Detection limit 1:32000 | |
NP_004455.2 | |
FGF5 | |
Peptide with sequence C-PLSAPRKNTNSVK corresponding to C-Terminus according to NP_004455.2, NP_149134.1. | |
0.1 mg | |
Angiogenesis, Cell Cycle and Replication | |
2250 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction