Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-8 Antibody (001), FITC , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290327F
Description
FGF-8 Monoclonal antibody specifically detects FGF-8 in Human samples. It is validated for ELISASpecifications
FGF-8 | |
Monoclonal | |
FITC | |
AIGFKAL6, Androgen-induced growth factor, FGF-8, fibroblast growth factor 8, fibroblast growth factor 8 (androgen-induced), HBGF-8, Heparin-binding growth factor 8, MGC149376 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human FGF-8 (NP_149355.1; Gln23-Arg204). | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
2253 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
001 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction