Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF-8A Recombinant Rabbit Monoclonal Antibody (HL2369)
GREENER_CHOICE

Catalog No. FEMA550507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550507 Supplier Invitrogen™ Supplier No. MA550507
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

FGF-8A Recombinant Monoclonal Antibody for IHC

FGF-8 is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8a, FGF-8b, FGF-8e and FGF-8f. The human and murine FGF-8a and FGF-8b are identical, unlike human and mouse FGF-8e and FGF-8f, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-8A
Applications Immunohistochemistry
Classification Recombinant Monoclonal
Clone HL2369
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene fgf8a
Gene Accession No. O57341
Gene Alias ace; cb110; etID309886.13; fb73a06; fgf8; fgf-8; ibd5031
Gene Symbols fgf8a
Host Species Rabbit
Immunogen Recombinant fragment of zebrafish Fgf8a.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30538
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.