Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-9 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299548B
Description
FGF-9 Polyclonal specifically detects FGF-9 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FGF-9 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse FGF-9. | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
2254 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction