Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-9 Antibody (FG9-77), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600623F
Description
FGF-9 Monoclonal specifically detects FGF-9 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry.Specifications
FGF-9 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FGF9 | |
Recombinant mouse FGF-9 | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
2254 | |
Human, Mouse | |
Purified |
ELISA, Immunohistochemistry | |
FG9-77 | |
ELISA, Immunohistochemistry | |
FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Monoclonal Anti-FGF9 reacts specifically with recombinant FGF9 (fibroblast growth factor 9). It does not cross-react with human FGF Acidic and FGF Basic. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction