Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ FGF-9, Mouse, Unlabeled, Clone: 3A10.1,

Catalog No. MABN1623 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1623 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1623 Supplier MilliporeSigma™ Supplier No. MABN1623
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fibroblast growth factor 9 (UniProt P31371; also known as FGF-9, GAF, Heparin-binding growth factor 9, Glia-activating factor, HBGF-9) is encoded by the FGF9 (also known as SYNS3) gene (Gene ID 2254) in human. Fibroblast growth factor 9 (FGF9) belongs to the FGF family of proteins that regulate a variety of cell functions, including proliferation, differentiation, survival, and motility, via FGF receptors (FGFRs). FGF-9 interacts with several FGFRs and binds preferentially to FGFR3. FGF-9 is highly expressed in cancer-associated fibroblasts (CAFs) in gastric cancer and may play a pivotal role in the interaction between cancer cells and their surrounding stromal cells. FGF-9 is shown to induce MMP-7 expression in AGS and MKN28 gastric cancer cells, promoting invasion and cellular anti-apoptotic activity through activation of the Erk and Akt signaling pathways. In human brain, FGF-9 is primarily expressed by neurons in the cortex, hippocampus, thalamus, cerebellum, and spinal cord, as well as by glia in the hindbrain and spinal cord. Upregulated FGF-9 expression is found in postmortem hippocampus tissues from major depressive disorder (MDD) patients, while FGF-2 is downregulated in the depressed brain. Likewise, chronic intracerebroventricular administration of FGF-9 increased both anxiety- and depression-like behaviors in rats, while FGF-9 knockdown in the dentate gyrus of the hippocampus ameliorated anxiety-like behavior. Human FGF-9 is produced with a propeptide (a.a. 1-3), which is cleaved off posttranslationally to yield the mature protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-9
Applications Western Blot
Classification Monoclonal
Clone 3A10.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2a in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene FGF9, SYNS3
Gene Accession No. P31371
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to a sequence within the C-terminal half of human FGF-9.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_002001
Test Specificity Clone 3A10.1 targets an internal sequence conserved among bovine, human, mouse, and rat species.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.