Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF acidic/FGF1 Antibody (V8P1G5*F2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250406AF700
Description
FGF acidic/FGF1 Monoclonal specifically detects FGF acidic/FGF1 in Zebrafish samples. It is validated for ELISA, Immunohistochemistry.Specifications
FGF acidic/FGF1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FGF1 | |
Ovalbumin-conjugated synthetic peptide LPRQVEQEED (C-terminal sequence) | |
0.1 mL | |
Angiogenesis, Cancer, Stem Cell Markers | |
2246 | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry | |
V8P1G5*F2 | |
ELISA, Immunohistochemistry | |
AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction