Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF acidic/FGF1 Antibody (V8P1G5*F2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | FGF acidic/FGF1 |
---|---|
Clone | V8P1G5*F2 |
Concentration | 1 mg/mL |
Dilution | ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | ELISA, Immunohistochemistry |
Description
FGF acidic/FGF1 Monoclonal antibody specifically detects FGF acidic/FGF1 in Zebrafish samples. It is validated for ELISA, ImmunohistochemistrySpecifications
FGF acidic/FGF1 | |
1 mg/mL | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Stem Cell Markers | |
PBS | |
2246 | |
IgG1 κ | |
Protein A purified |
V8P1G5*F2 | |
ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Zebrafish | |
AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 | |
Ovalbumin-conjugated synthetic peptide LPRQVEQEED (C-terminal sequence) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title