Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF acidic/FGF1 Rabbit anti-Human, Mouse, Rat, Clone: 3X3E5, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316609100UL
This item is not returnable.
View return policy
Description
FGF acidic/FGF1 Monoclonal antibody specifically detects FGF acidic/FGF1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
FGF acidic/FGF1 | |
Monoclonal | |
Unconjugated | |
AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 | |
A synthetic peptide corresponding to a sequence within amino acids 56-155 of human FGF acidic/FGF1 (P05230). | |
100 μg | |
Angiogenesis, Biologically Active Proteins, Cancer, Stem Cell Markers | |
2246 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
3X3E5 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction