Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody (013), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289192F
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FGF basic/FGF2/bFGF | |
Monoclonal | |
FITC | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
This antibody was obtained from a rabbit immunized with purified, synthetic peptide corresponding to the N-terminus of the Human FGF basic/FGF2/bFGF. | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer | |
2247 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
13 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction