Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody (FB-8), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001128AF647
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human, Bovine samples. It is validated for Western Blot, ELISA.Specifications
FGF basic/FGF2 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FGF2 | |
Recombinant full length protein (Human). | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer | |
2247 | |
Human, Bovine | |
Purified |
Western Blot, ELISA | |
FB-8 | |
Western Blot, ELISA | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not cross-react with human fibroblast growth factor - acidic (aFGF). | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction