Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF basic/FGF2/bFGF Antibody (MC-GF1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$487.50

Specifications

Antigen FGF basic/FGF2
Clone MC-GF1
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600778
SDP
View Documents
Novus Biologicals
NB600778
0.25 mg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

FGF basic/FGF2
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Purified
RUO
Human
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
FGF2
IgG2a
Protein A or G purified
GF1 recognizes basic fibroblast growth factor (bFGF). The binds strongly to bFGF attached to proteoglycans, but is competitively displaced by heparin.
MC-GF1
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Angiogenesis, Cancer
P09038
2247
Recombinant full length protein (Human).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.