Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-BP Antibody (OTI2B6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271527IR
Description
FGF-BP Monoclonal specifically detects FGF-BP in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FGF-BP | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FGFBP1 | |
Recombinant protein fragment corresponding to amino acids 24-234 of human FGF-BP (NP_005121) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2B6 | |
Western Blot, Immunohistochemistry | |
17 kDa heparin-binding growth factor-binding protein, FGF-BP, FGF-BP1, FGFBP-1, FGFBPFGF-binding protein 1, fibroblast growth factor binding protein 1, fibroblast growth factor-binding protein 1, HBp17, HBP1717 kDa HBGF-binding protein, heparin-binding growth factor binding protein | |
Mouse | |
Affinity Purified | |
Angiogenesis, Cell Cycle and Replication | |
9982.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction