Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF Receptor-beta Monoclonal Antibody (M17A3), QED Bioscience™
SDP

Catalog No. 502277946
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7946 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7946 Supplier Leinco Technologies Inc Supplier No. 30106250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF Receptor-beta Monoclonal antibody specifically detects FGF Receptor-beta in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen FGF Receptor-beta
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M17A3
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FGFR1
Gene Accession No. P11362
Host Species Mouse
Immunogen Recombinant human ectodomain of FGFr1 beta expressed in E. coli beginning with pro23; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2260
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.