Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF10 Monoclonal Antibody (04), Invitrogen™

Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | FGF10 |
---|---|
Clone | 04 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543398
|
Invitrogen™
MA543398 |
50 μL |
Each for $285.50
|
|
|||||
PIMA542993
![]() |
Invitrogen™
MA542993 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human FGF10/KGF2
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.Specifications
FGF10 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria | |
FGF10 | |
IgG2a | |
Protein A | |
Antibody |
04 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
O15520 | |
2255 | |
Recombinant Human FGF10/KGF2 Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
FGF10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title