Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103408 Supplier Invitrogen™ Supplier No. PA5103408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FGF11.

Probably involved in nervous system development and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FGF11
Gene Accession No. P70378, Q92914
Gene Alias FGF; Fgf11; Fgf-11; Fhf3; FHF-3; Fibroblast growth factor; Fibroblast growth factor 11; fibroblast growth factor homologous factor 3
Gene Symbols FGF11
Host Species Rabbit
Immunogen A synthesized peptide derived from human FGF11(Accession Q92914), corresponding to amino acid residues P66-I116.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14166, 170632, 2256
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.