Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF11 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA566853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566853 Supplier Thermo Scientific Supplier No. PA566853

Rabbit Polyclonal Antibody

Immunogen sequence: VTKLFCRQGFY LQANPDGSIQ GTPEDTSSFT HFN Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Probably involved in nervous system development and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF11
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FGF11
Gene Accession No. Q92914
Gene Alias FGF; Fgf11; Fgf-11; Fhf3; FHF-3; Fibroblast growth factor; Fibroblast growth factor 11; fibroblast growth factor homologous factor 3
Gene Symbols FGF11
Host Species Rabbit
Immunogen Recombinant Human FGF11
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2256
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.