Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF13 Monoclonal Antibody (OTI7E8)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526916

Catalog No. PIMA526916


Only null left
Add to Cart

Description

Description

FGF13 Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This gene is located in a region on chromosome X, which is associated with Borjeson-Forssman-Lehmann syndrome (BFLS), making it a possible candidate gene for familial cases of the BFLS, and for other syndromal and nonspecific forms of X-linked mental retardation mapping to this region. Alternative splicing of this gene at the 5' end results in several transcript variants encoding different isoforms with different N-termini.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FGF13
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q92913
FGF13
Full length human recombinant protein of FGF13 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI7E8
Unconjugated
FGF13
AFGF; ECGF; ECGFA; ECGFB; FGF; FGF 1; FGF alpha; FGF13; FGF-13; FGF13A; FGF2; FGFA; Fhf2; FHF-2; Fibroblast growth factor; fibroblast growth factor 13; Fibroblast growth factor homologous factor 2; GLIO703; HBGF 1; HBGF1
Mouse
Affinity Chromatography
RUO
2258
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.