Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGF14 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537861
Description
This antibody is predicted to react with rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
FGF14 is involved in nervous system development and function.
Specifications
| FGF14 | |
| Polyclonal | |
| Unconjugated | |
| FGF14 | |
| bA397O8.2; FGF; FGF homologus factor 4b; Fgf14; FGF-14; FHF4; FHF-4; Fibroblast growth factor; Fibroblast growth factor 14; fibroblast growth factor 14, isoform b; fibroblast growth factor homologous factor 4; m 1B; mFHF-4(1B); SCA27 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 14169, 2259 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P70379, Q92915 | |
| FGF14 | |
| Peptide with sequence REQHWDRPSASR-C, from the N-terminus of mouse Fgf14 (aa 17-28). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction