Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577979 Supplier Invitrogen™ Supplier No. PA577979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Mouse (98%), Rat (95%), Zebrafish (87%), Chicken (94%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

FGF14 is involved in nervous system development and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF14
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene FGF14
Gene Accession No. Q92915
Gene Alias bA397O8.2; FGF; FGF homologus factor 4b; Fgf14; FGF-14; FHF4; FHF-4; Fibroblast growth factor; Fibroblast growth factor 14; fibroblast growth factor 14, isoform b; fibroblast growth factor homologous factor 4; m 1B; mFHF-4(1B); SCA27
Gene Symbols FGF14
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FGF14. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2259
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.