Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF14 Rabbit anti-Human, Clone: 107, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290235100UL
Description
FGF14 Monoclonal specifically detects FGF14 in Human samples. It is validated for ELISA.Specifications
FGF14 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
107 | |
ELISA 1:5000-1:10000 | |
bA397O8.2, FGF-14, FHF4FHF-4, fibroblast growth factor 14, Fibroblast growth factor homologous factor 4, SCA27MGC119129 | |
Recombinant Human FGF14 protein | |
100 μg | |
Angiogenesis, Neuroscience | |
2259 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction