Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGF16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550664
Description
The antibody detects endogenous levels of total FGF16 protein.
FGF-16 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system.
Specifications
FGF16 | |
Polyclonal | |
Unconjugated | |
FGF16 | |
FGF; FGF16; FGF-16; Fibroblast growth factor; Fibroblast growth factor 16; metacarpal 4-5 fusion; MF4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60464, 80903, 8823 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O43320, O54769, Q9ESL8 | |
FGF16 | |
Synthetic peptide corresponding to a region derived from internal residues of human fibroblast growth factor 16. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction