Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF18 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306688
This item is not returnable.
View return policy
Description
FGF18 Polyclonal antibody specifically detects FGF18 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISASpecifications
FGF18 | |
Polyclonal | |
Unconjugated | |
PBS (without Mg2+, Ca2+) (pH 7.4), 150 mM NaCl, 50% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA | |
1 mg/ml | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:100, Peptide ELISA 1:40000 | |
FGF-18ZFGF5, fibroblast growth factor 18, zFGF5 | |
Synthetic peptide from human FGF18. (AA Range: 151-200) | |
50 μg | |
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
8817 | |
Store at -20°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction