Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF18 Mouse anti-Human, Mouse, Clone: 1D11D12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | FGF18 |
---|---|
Clone | 1D11D12 |
Concentration | 0.74 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6428
![]() ![]() |
Proteintech Group Inc
603411IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
FGF18 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. It has been shown in vitro that this protein is able to induce neurite outgrowth in PC12 cells. Studies of the similar proteins in mouse and chick suggested that this protein is a pleiotropic growth factor that stimulates proliferation in a number of tissues, most notably the liver and small intestine. Knockout studies of the similar gene in mice implied the role of this protein in regulating proliferation and differentiation of midline cerebellar structures.Specifications
FGF18 | |
0.74 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FGF 18, FGF18, fibroblast growth factor 18, ZFGF5 | |
FGF18 | |
IgG2a | |
Protein A | |
Antibody |
1D11D12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O76093, O89101 | |
14172, 8817 | |
FGF18 Fusion Protein Ag2041 | |
Primary | |
-20°C | |
FGF18 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title