Learn More
FGF19 Monoclonal Antibody (117601), Invitrogen™
Description
In Western blots, no cross-reactivity with recombinant human (rh) FGF acidic, rhFGF basic, rhFGF-4, -5, -6, -7, -9, -10, -17, -18, recombinant mouse FGF-8b, -8c, or -15 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | FGF19 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 117601 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | FGF19 |
| Gene Accession No. | O95750 |
| Gene Alias | FGF; Fgf15; FGF19; FGF-19; Fibroblast growth factor; fibroblast growth factor 15; fibroblast growth factor 19; UNQ334/PRO533 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.