Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF19 Monoclonal Antibody (117611)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA523861

Catalog No. PIMA523861


Only null left
Add to Cart

Description

Description

This antibody shows approximately 10% cross-reactivity with recombinant human (rh) FGF-3 and rhFGF-5 and no cross-reactivity with rhFGF-4, rhFGF-6, rhFGF-7, rhFGF-9, rhFGF-10, rhFGF-13, rhFGF-16, rhFGF-17, rhFGF-18, rhFGF acidic, and rhFGF basic. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

FGF19, Fibroblast growth factor 19, is a protein that in humans is encoded by the FGF19 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. The FGF19 gene is mapped to 11q13.3. The deduced 216-amino acid FGF19 protein contains asignal sequence and 2 cysteine residues that are conserved in the FGF family. Expression of this gene was detected only in fetal but not adult brain tissue. Synergistic interaction of the chick homolog and Wnt-8c has been shown to be required for initiation of inner ear development. FGF19 stimulates hepatic protein and glycogen synthesis but does not induce lipogenesis. The effects of FGF19 are independent of the activity of either insulin or the protein kinase Akt and, instead, are mediated through a mitogen-activated protein kinase signaling pathway that activates components of the protein translation machinery and stimulates glycogen synthase activity. The orthologous protein in mouse is FGF15, which shares about 50% amino acid identity and has similar functions. Together they are often referred to as FGF15/19.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FGF19
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
O95750
FGF19
E. coli-derived recombinant human FGF-19 Phe27-Lys216.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Neutralization, Western Blot
117611
Unconjugated
FGF19
FGF; Fgf15; FGF19; FGF-19; Fibroblast growth factor; fibroblast growth factor 15; fibroblast growth factor 19; UNQ334/PRO533
Mouse
Protein A/G
RUO
9965
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.