Learn More
Invitrogen™ FGF2 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
$459.00 - $1159.00
Specifications
Antigen | FGF2 Biosimilar |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
Classification | Recombinant Monoclonal |
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term HuGAL-F2.
FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.Specifications
FGF2 Biosimilar | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Lyophilized | |
IgG2SA | |
Human | |
Primary | |
Protein A |
1 mg/mL | |
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
Unconjugated | |
Human | |
RUO | |
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.