Learn More
Invitrogen™ FGF2 Monoclonal Antibody (10060)

Description
In capture ELISAs, this antibody shows approximately 0.2% cross-reactivity with bovine FGF acidic, and no cross-reactivity with rhFGF acidic, rhFGF-4, rhFGF-6, or rhFGF-7. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | FGF2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 10060 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | FGF2 |
| Gene Accession No. | P09038 |
| Gene Alias | Basic fibroblast growth factor; basic fibroblast growth factor bFGF; bFGF; FGF; fgf basic; Fgf2; Fgf-2; Fgfb; FGF-b; Fibroblast growth factor; fibroblast growth factor 2; fibroblast growth factor 2 (basic); Fibroblast growth factor basic; HBGF-2; Heparin-binding growth factor 2; H-FGF-b-147; H-FGF-b-154; M-FGF-b; Prostatic growth factor; prostatropin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.