Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF21 Monoclonal Antibody (OTI1B4), TrueMAB™, OriGene
SDP

Catalog No. 502541311
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1311 30 μL
50-167-3800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1311 Supplier OriGene Supplier No. TA502787S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF21 Monoclonal antibody specifically detects FGF21 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen FGF21
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1B4
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FGF21
Gene Accession No. Q9NSA1
Gene Alias FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 29-209 of human FGF21 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26291
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.