Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGF21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115227
Description
Antibody detects endogenous levels of total FGF21.
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
Specifications
| FGF21 | |
| Polyclonal | |
| Unconjugated | |
| FGF21 | |
| FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26291, 56636 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9JJN1, Q9NSA1 | |
| FGF21 | |
| A synthesized peptide derived from human FGF21(Accession Q9NSA1), corresponding to amino acid residues A162-S209. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction