Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120088 Supplier Invitrogen™ Supplier No. PA5120088
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: LO2, Mouse pancreas. The target is usually found in the following locations: Secreted. Immunogen sequence: HPIPDSSPLL QFGGQVRQRY LYTDDAQQTE AHLEIREDGT VGGAADQSPE SLLQLKALKP GVIQILGVKT SRFLCQRPDG ALYGSLHFDP EACSFRELLL EDGYNVYQSE AHGLPLHLPG NKSPHRDPAP RGPARFLPLP GLPPALPEPP GILAPQPPDV GSSDPLSMVG PSQGRSPSYA S.

The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF21
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FGF21
Gene Accession No. Q9JJN1, Q9NSA1
Gene Alias FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196
Gene Symbols FGF21
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 29-209 of human FGF21 (NP_061986.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26291, 56636
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.