Learn More
Invitrogen™ FGF22 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA550960
Catalog No. PA550960
Only null left
Add to Cart
Description
Description
FGF22 Polyclonal Antibody detects endogenous levels of FGF22 protein.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The mouse homolog of this gene was found to be preferentially expressed in the inner root sheath of the hair follicle, which suggested a role in hair development. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF22
Polyclonal
Unconjugated
FGF22
2210414E06Rik; FGF; Fgf22; FGF-22; Fibroblast growth factor; Fibroblast growth factor 22; UNQ2500/PRO5800
Rabbit
Antigen affinity chromatography
RUO
170579, 27006, 67112
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Q9ESS2, Q9HCT0
FGF22
Synthesized peptide derived from the Internal region of human FGF-22 (aa 71-120).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.