Learn More
Invitrogen™ FGF22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA594992
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human Caco-2 whole cell, human U-87MG whole cell, human PC-3 whole cell, rat brain tissue, rat testicular tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The mouse homolog of this gene was found to be preferentially expressed in the inner root sheath of the hair follicle, which suggested a role in hair development. Alternative splicing results in multiple transcript variants.
Specifications
FGF22 | |
Polyclonal | |
Unconjugated | |
FGF22 | |
2210414E06Rik; FGF; Fgf22; FGF-22; Fibroblast growth factor; Fibroblast growth factor 22; UNQ2500/PRO5800 | |
Rabbit | |
Affinity chromatography | |
RUO | |
170579, 27006, 67112 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
Q9ESS2, Q9HCT0 | |
FGF22 | |
A synthetic peptide corresponding to a sequence in the middle region of human FGF22 (93-109aa FYVAMNRRGRLYGSRLY). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.