Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF23, Mouse anti-Human, Clone: FG322-3, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16059
Specifications
FGF23 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4. | |
ADHR/HPDR2/HYPF/PHPTC | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
FG322-3 | |
ELISA (1:2000-5000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-500) Western Blot (1:2000-5000) The optimal working dilution should be determined by the end user. | |
fibroblast growth factor 23 | |
FGF23 | |
Recombinant protein corresponding to human FGF23. | |
RUO | |
8074 | |
For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction