Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597747
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597747 Supplier Invitrogen™ Supplier No. PA597747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FGF23 Polyclonal Antibody for Western Blot, ELISA

The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF23
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FGF23
Gene Accession No. Q8VI82
Gene Alias ADHR; FGF; Fgf23; FGF-23; FGFN; Fibroblast growth factor; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 N-terminal peptide; HPDR2; HYPF; Phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor; UNQ3027/PRO9828
Gene Symbols FGF23
Host Species Rabbit
Immunogen Recombinant Rat Fibroblast growth factor 23 protein (25-251AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170583
Target Species Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.