Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF3 Monoclonal Antibody (MSD1)
GREENER_CHOICE

Catalog No. PIMA146094
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA146094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA146094 Supplier Invitrogen™ Supplier No. MA146094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF3 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, IP

Fibroblast growth factors (FGFs) can stimulate the proliferation of mesenchymal, epithelial and neuroectodermal cells. FGF3 belongs to the heparin binding growth factors family. Members of the family include Acidic FGF (also known as aFGF or FGF 1), basic FGF (bFGF or FGF 2), FGF 3 (also known as Int2) and FGF 4 (also known as hst or Kaposi FGF). FGF 4 was first revealed to be an oncoprotein in Kaposi's sarcoma cells. Other family members include FGF 5, FGF 6, FGF 7 (keratinocyte growth factor, KGF), FGF 8 (AIGF), FGF 9 (GAF) and FGF 10. FGF3 is activated by proviral insertion of mouse mammary tumor virus (MMTV). FGF3 is responsible for most of breast malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MSD1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FGF3
Gene Accession No. P05524, P11487, P36386
Gene Alias FGF; fgf3; FGF-3; fgf3.L; fgf3-A; Fibroblast growth factor; Fibroblast growth factor 3; fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog); fibroblast growth factor 3 L homeolog; Fibroblast growth factor-3; HBGF-3; Heparin-binding growth factor 3; id:ibd5006; INT2; int-2; INT-2 proto-oncogene protein; interacting gene 2; Int-P; lia; murine mammary tumor virus integration site 2, mouse; oncogene INT2; proto-oncogene Int-2; V-INT2 murine mammary tumor virus integration site oncogene homolog; XELAEV_18021863mg; xfgf3; xfgf-3
Gene Symbols FGF3, fgf3.L
Host Species Mouse
Immunogen Peptide conjugated to BSA.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14174, 2248, 373669
Target Species Human, Mouse, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.