Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552971
Description
Immunogen sequence: RGRLYASEHY SAECEFVERI HELGYNTYAS RLYRTVSSTP GARRQPSAER LWYVSVNGKG RPRRGFKTRR TQKSSLFLPR VLDHRDHEMV RQLQSGLPRP PGKGVQPRRR RQKQSPDNLE PS Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
Fibroblast growth factors (FGFs) can stimulate the proliferation of mesenchymal, epithelial and neuroectodermal cells. FGF3 belongs to the heparin binding growth factors family. Members of the family include Acidic FGF (also known as aFGF or FGF 1), basic FGF (bFGF or FGF 2), FGF 3 (also known as Int2) and FGF 4 (also known as hst or Kaposi FGF). FGF 4 was first revealed to be an oncoprotein in Kaposi's sarcoma cells. Other family members include FGF 5, FGF 6, FGF 7 (keratinocyte growth factor, KGF), FGF 8 (AIGF), FGF 9 (GAF) and FGF 10. FGF3 is activated by proviral insertion of mouse mammary tumor virus (MMTV). FGF3 is responsible for most of breast malignancies.
Specifications
| FGF3 | |
| Polyclonal | |
| Unconjugated | |
| FGF3 | |
| FGF; fgf3; FGF-3; fgf3.L; fgf3-A; Fibroblast growth factor; Fibroblast growth factor 3; fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog); fibroblast growth factor 3 L homeolog; Fibroblast growth factor-3; HBGF-3; Heparin-binding growth factor 3; id:ibd5006; INT2; int-2; INT-2 proto-oncogene protein; interacting gene 2; Int-P; lia; murine mammary tumor virus integration site 2, mouse; oncogene INT2; proto-oncogene Int-2; V-INT2 murine mammary tumor virus integration site oncogene homolog; XELAEV_18021863mg; xfgf3; xfgf-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2248 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11487 | |
| FGF3 | |
| Recombinant protein corresponding to Human FGF3. Recombinant protein control fragment (Product #RP-89880). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction