Learn More
Invitrogen™ FGF4 Polyclonal Antibody

Description
Immunogen sequence: APTAPNGTLE AELERRWESL VALSLARLPV AAQPKEAAVQ SGAGDYLLGI KRLRRLYCNV GIGFHLQALP DGRIGGAHAD TRDSLLELSP VERGVVSIFG VASRFFVAMS SKGKLYGSPF FTDECTFKEI LLPNNYNAYE SYKYPGMFIA LSKNGKTKKG NRVSPTMKVT HFLPRL; Positive Samples: Rat liver.
Specifications
Specifications
| Antigen | FGF4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Fgf4 |
| Gene Alias | FGF; FGF4; Fgf-4; Fgfk; Fibroblast growth factor; fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; heparin secretory-transforming protein 1; heparin-binding growth factor 1; heparin-binding growth factor 4; HST; Hst1; hst-1; HSTF1; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; kaposi sarcoma oncogene; Kfgf; K-FGF; K-fibroblast growth factor; KS3; oncogene HST; transforming protein KS3 |
| Gene Symbols | Fgf4 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.