Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGF8 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537860
Description
This antibody is predicted to react with mouse, rat, dog and pig based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. Required for normal brain, eye, ear and limb development during embryogenesis. Required for normal development of the gonadotropin-releasing hormone (GnRH) neuronal system. Plays a role in neurite outgrowth in hippocampal cells.
Specifications
FGF8 | |
Polyclonal | |
Unconjugated | |
FGF8 | |
AIGF; androgen-induced; Androgen-induced growth factor; FGF; FGF8; Fgf-8; Fibroblast growth factor; Fibroblast growth factor 8; fibroblast growth factor 8 (androgen-induced); HBGF-8; Heparin-binding growth factor 8; HH6; KAL6; MGC149376 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
2253 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P55075 | |
FGF8 | |
Peptide with sequence C-HTTEQSLRFEFLNYP, from the internal region of FGF8 (aa 201-216). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction