Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGFBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596685 Supplier Invitrogen™ Supplier No. PA596685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKKVKNGLHS KVVSEQKDTL GNTQIKQKSR PGNKGKFVTK DQANCRWAAT EQEEGISLKV ECTQLDHEFS CVFAGNPTSC LKLKDERVYW KQVARNLRSQ KDICRYSKTA VKTRVCRKDF PESSLKLVSS TLFGNTKPRK EKTEMSPREH IKGKETTPSS LAVTQTMATK APECVEDPDM ANQRKTALEF CGETWSSLCT FFLSIVQDTS C; Positive Samples: A-549, HepG2, NIH/3T3, Mouse kidney, Mouse lung, Rat brain; Cellular Location: Cell membrane, Peripheral membrane protein, extracellular space.

FGFBP1 encodes a secreted fibroblast growth factor carrier protein. The encoded protein plays a critical role in cell proliferation, differentiation and migration by binding to fibroblast growth factors and potentiating their biological effects on target cells. The encoded protein may also play a role in tumor growth as an angiogenic switch molecule, and expression of this gene has been associated with several types of cancer including pancreatic and colorectal adenocarcinoma. A pseudogene of this gene is also located on the short arm of chromosome 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFBP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FGFBP1
Gene Accession No. O70514, Q14512, Q9QY10
Gene Alias 17 kDa HBGF-binding protein; 17 kDa heparin-binding growth factor-binding protein; FGF-binding protein 1; FGFBP; FGF-BP; Fgfbp1; FGF-BP1; FGFBP-1; fibroblast growth factor binding protein 1; fibroblast growth factor-binding protein 1; growth factor binding protein-1; growth factor-binding protein 1; HBP17; heparin-binding growth factor binding protein
Gene Symbols FGFBP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-234 of human FGFBP1 (NP_0051211).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14181, 64535, 9982
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.