Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFBP2/KSP37 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981234
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1234 Supplier Bioss Supplier No. BS11284R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the fibroblast growth factor binding protein family. The encoded protein is a serum protein that is selectively secreted by cytotoxic lymphocytes and may be involved in cytotoxic lymphocyte-mediated immunity. An increase in the amount of gene product may be associated with atopic asthma and mild extrinsic asthma.

Specifications

Antigen FGFBP2/KSP37
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FGFBP2
Gene Accession No. Q9BYJ0
Gene Alias 37 kDa killer-specific secretory protein; FGF-binding protein 2; FGFBP2; FGF-BP2; FGFBP-2; fibroblast growth factor binding protein 2; Fibroblast growth factor-binding protein 2; HBp17-related protein; HBP17RP; HBp17-RP; killer-specific secretory protein of 37 kDa; KSP37; UNQ425/PRO1065
Gene Symbols FGFBP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FGFBP2/KSP37.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83888
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.