Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFBP3 Antibody (002), HRP , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289733H
Description
FGFBP3 Monoclonal antibody specifically detects FGFBP3 in Human samples. It is validated for ELISASpecifications
FGFBP3 | |
Monoclonal | |
HRP | |
C10orf13, Chromosome 10 Open Reading Frame 13, FGF-Binding Protein 3, FGFBP3, FGF-BP3, FGFBP-3, Fibroblast Growth Factor-Binding Protein 3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human FGFBP3 (Q8TAT2; Met1-Gly258). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
002 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
143282 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction