Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGFBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103412 Supplier Invitrogen™ Supplier No. PA5103412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FGFBP3.

FGFBP3 is a heparin-binding protein which binds to FGF2, prevents binding of FGF2 to heparin and probably inhibits immobilization of FGF2 on extracellular matrix glycosaminoglycans, allowing its release and subsequent activation of FGFR signaling which leads to increased vascular permeability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFBP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FGFBP3
Gene Accession No. Q8TAT2
Gene Alias 2610306H15Rik; C10orf13; FGF-binding protein 3; Fgfbp3; Fgf-bp3; FGFBP-3; fibroblast growth factor binding protein 3; fibroblast growth factor-binding protein 3; PSEC0101
Gene Symbols FGFBP3
Host Species Rabbit
Immunogen A synthesized peptide derived from human FGFBP3(Accession Q8TAT2), corresponding to amino acid residues E201-F251.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143282
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.